Learn More
Biomatik Corporation Human Glucose Transporter 1 (GLUT1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04413
The Human Glucose Transporter 1 (GLUT1) ELISA Kit allows for quantitative determination of Glucose Transporter 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.116 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P11166. Cross Activity: No significant cross-reactivity or interference between Glucose Transporter 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2590