Learn More
Biomatik Corporation Human Interleukin 12A (IL12A) ELISA Kit
Supplier:  Biomatik Corporation EKU05201
Encompass_Preferred
Human Interleukin 12A (IL12A) ELISA Kit for detecting IL12A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29459. Gene ID: 3592. Target Synonyms: p35; IL12-A; CLMF1; NFSK; NKSF1; Natural Killer Cell Stimulatory Factor 1; Cytotoxic Lymphocyte Maturation Factor 1; NF Cell Stimulatory Factor Chain 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6128
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.