Learn More
Biomatik Corporation Mouse Glutaredoxin (GLRX) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04471
The Mouse Glutaredoxin (GLRX) ELISA Kit allows for quantitative determination of Glutaredoxin in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 78-5000 pg/mL. Sensitivity: 35 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9QUH0. Cross Activity: No significant cross-reactivity or interference between Glutaredoxin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2615