Learn More
Biomatik Corporation Glutathione (GSH) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04473
The Glutathione (GSH) ELISA Kit allows for quantitative determination of Glutathione in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: All species. Detection Range: 1.23-100 ug/mL. Sensitivity: 0.45 ug/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Glutathione and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2617