Learn More
Biomatik Corporation Human Heat Shock 60kD Protein 1, Chaperonin (HSPD1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04647
The Human Heat Shock 60kD Protein 1, Chaperonin (HSPD1) ELISA Kit allows for quantitative determination of Heat Shock 60kD Protein 1, Chaperonin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.64 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P10809. Cross Activity: No significant cross-reactivity or interference between Heat Shock 60kD Protein 1, Chaperonin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2687