Learn More
Biomatik Corporation Human Hemoglobin Beta (HBb) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04721
The Human Hemoglobin Beta (HBb) ELISA Kit allows for quantitative determination of Hemoglobin Beta in Serum, Plasma and Erythrocyte lysates. Species Reactivity: Homo sapiens (Human). Detection Range: 6.17-500 ug/mL. Sensitivity: 2.94 ug/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P68871. Cross Activity: No significant cross-reactivity or interference between Hemoglobin Beta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2711