Learn More
Biomatik Corporation Mouse Hepatocyte Growth Factor (HGF) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04752
The Mouse Hepatocyte Growth Factor (HGF) ELISA Kit allows for quantitative determination of Hepatocyte Growth Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q08048. Cross Activity: No significant cross-reactivity or interference between Hepatocyte Growth Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2724