Learn More
Biomatik Corporation Cattle Hepcidin (Hepc) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04762
The Cattle Hepcidin (Hepc) ELISA Kit allows for quantitative determination of Hepcidin in Serum, Plasma and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.214 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q2NKT0. Cross Activity: No significant cross-reactivity or interference between Hepcidin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2729