Learn More
Biomatik Corporation Rat Hypoxia Inducible Factor 1 Alpha (HIF1a) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04863
The Rat Hypoxia Inducible Factor 1 Alpha (HIF1a) ELISA Kit allows for quantitative determination of Hypoxia Inducible Factor 1 Alpha in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O35800. Cross Activity: No significant cross-reactivity or interference between Hypoxia Inducible Factor 1 Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2766