Learn More
Biomatik Corporation Mouse Hypoxia Inducible Factor 2 Alpha (HIF2a) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04867
The Mouse Hypoxia Inducible Factor 2 Alpha (HIF2a) ELISA Kit allows for quantitative determination of Hypoxia Inducible Factor 2 Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 78-5,000 pg/mL. Sensitivity: 34 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P97481. Cross Activity: No significant cross-reactivity or interference between Hypoxia Inducible Factor 2 Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2768