Learn More
Biomatik Corporation Rat Immunoglobulin E (IgE) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04888
The Rat Immunoglobulin E (IgE) ELISA Kit allows for quantitative determination of Immunoglobulin E in Serum, Plasma, and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 31.2-2000 pg/mL. Sensitivity: 14.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01855. Cross Activity: No significant cross-reactivity or interference between Immunoglobulin E and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2781