Learn More
Biomatik Corporation Human Immunoglobulin M (IgM) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04911
The Human Immunoglobulin M (IgM) ELISA Kit allows for quantitative determination of Immunoglobulin M in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 ng/mL. Sensitivity: 13.2 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. Cross Activity: No significant cross-reactivity or interference between Immunoglobulin M and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2786