Learn More
Biomatik Corporation Cattle Insulin (INS) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04954
The Cattle Insulin (INS) ELISA Kit allows for quantitative determination insulin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 123.5-10000 pg/mL. Sensitivity: 47.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01317. Cross Activity: No significant cross-reactivity or interference between Insulin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2799