Learn More
Biomatik Corporation Mouse Insulin Like Growth Factor Binding Protein 3 (IGFBP3) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04995
The Mouse Insulin Like Growth Factor Binding Protein 3 (IGFBP3) ELISA Kit allows for quantitative determination insulin Like Growth Factor Binding Protein 3 in Serum, Plasma and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.94-60 ng/mL. Sensitivity: 0.37 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P47878. Cross Activity: No significant cross-reactivity or interference between Insulin Like Growth Factor Binding Protein 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2819