Learn More
Biomatik Corporation Human Insulin Like Growth Factor Binding Protein 6 (IGFBP6) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU05006
The Human Insulin Like Growth Factor Binding Protein 6 (IGFBP6) ELISA Kit allows for quantitative determination insulin Like Growth Factor Binding Protein 6 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.064 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P24592. Cross Activity: No significant cross-reactivity or interference between Insulin Like Growth Factor Binding Protein 6 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2826