Learn More
Biomatik Corporation Human Intercellular Adhesion Molecule 5 (ICAM5) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU05067
The Human Intercellular Adhesion Molecule 5 (ICAM5) ELISA Kit allows for quantitative determination intercellular Adhesion Molecule 5 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UMF0. Cross Activity: No significant cross-reactivity or interference between Intercellular Adhesion Molecule 5 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2855