Learn More
Biomatik Corporation Gallus Interferon Alpha (IFNa) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU05072
The Gallus Interferon Alpha (IFNa) ELISA Kit allows for quantitative determination interferon Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P42165. Cross Activity: No significant cross-reactivity or interference between Interferon Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2858