Learn More
Biomatik Corporation Mouse Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU05157
The Mouse Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit allows for quantitative determination interleukin 1 Receptor Antagonist in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.12-200 pg/mL. Sensitivity: 1.33 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25085. Cross Activity: No significant cross-reactivity or interference between Interleukin 1 Receptor Antagonist and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2911