Learn More
Biomatik Corporation Mouse Interleukin 1 Receptor Type II (IL1R2) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU05169
The Mouse Interleukin 1 Receptor Type II (IL1R2) ELISA Kit allows for quantitative determination interleukin 1 Receptor Type II in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.62-1000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P27931. Cross Activity: No significant cross-reactivity or interference between Interleukin 1 Receptor Type II and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2913