Learn More
Biomatik Corporation Mouse Interleukin 11 Receptor Alpha (IL11Ra) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU05191
The Mouse Interleukin 11 Receptor Alpha (IL11Ra) ELISA Kit allows for quantitative determination interleukin 11 Receptor Alpha in Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.061 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q64385. Cross Activity: No significant cross-reactivity or interference between Interleukin 11 Receptor Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2929