Learn More
Biomatik Corporation Human Interleukin 23 (IL23) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU05295
The Human Interleukin 23 (IL23) ELISA Kit allows for quantitative determination interleukin 23 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 2.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NPF7. Cross Activity: No significant cross-reactivity or interference between Interleukin 23 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2982