Learn More
Biomatik Corporation Rabbit Interleukin 4 (IL4) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU05339
The Rabbit Interleukin 4 (IL4) ELISA Kit allows for quantitative determination interleukin 4 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9MZR8. Cross Activity: No significant cross-reactivity or interference between Interleukin 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3012