Learn More
Biomatik Corporation Rat JNK1/MAPK8 Associated Membrane Protein (JKAMP) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU05418
The Rat JNK1/MAPK8 Associated Membrane Protein (JKAMP) ELISA Kit allows for quantitative determination of JNK1/MAPK8 Associated Membrane Protein in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.55 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: D3ZZT8. Cross Activity: No significant cross-reactivity or interference between JNK1/MAPK8 Associated Membrane Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3052