Learn More
Biomatik Corporation Mouse Kallikrein 6 (KLK6) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU05446
The Mouse Kallikrein 6 (KLK6) ELISA Kit allows for quantitative determination of Kallikrein 6 in Serum, Plasma, Cerebrospinal fluid, Breast milk and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.2-2000 pg/mL. Sensitivity: 13.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P15947. Cross Activity: No significant cross-reactivity or interference between Kallikrein 6 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3061