Learn More
Biomatik Corporation Rat Keratin 18 (KRT18) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05469
Encompass_Preferred
The Rat Keratin 18 (KRT18) ELISA Kit allows for quantitative determination of Keratin 18 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q5BJY9. Cross Activity: No significant cross-reactivity or interference between Keratin 18 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3071