Learn More
Biomatik Corporation Mouse Ki-67 Protein (Ki67P) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU05486
The Mouse Ki-67 Protein (Ki67P) ELISA Kit allows for quantitative determination of Ki-67 Protein in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.122 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: E9PVX6. Cross Activity: No significant cross-reactivity or interference between Ki-67 Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3079