Learn More
Biomatik Corporation Human Laminin Receptor 1 (LAMR1) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU05563
The Human Laminin Receptor 1 (LAMR1) ELISA Kit allows for quantitative determination of Laminin Receptor 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 27 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08865. Cross Activity: No significant cross-reactivity or interference between Laminin Receptor 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3105