Learn More
Biomatik Corporation Mouse Lecithin Cholesterol Acyltransferase (LCAT) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU05571
The Mouse Lecithin Cholesterol Acyltransferase (LCAT) ELISA Kit allows for quantitative determination of Lecithin Cholesterol Acyltransferase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.062 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P16301. Cross Activity: No significant cross-reactivity or interference between Lecithin Cholesterol Acyltransferase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3109