Learn More
Biomatik Corporation Rat Macrophage Derived Chemokine (MDC) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU05742
The Rat Macrophage Derived Chemokine (MDC) ELISA Kit allows for quantitative determination of Macrophage Derived Chemokine in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 23.44-1500 pg/mL. Sensitivity: 8.47 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q91ZH5. Cross Activity: No significant cross-reactivity or interference between Macrophage Derived Chemokine and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3179