Learn More
Biomatik Corporation Human Major Histocompatibility Complex Class I A (MHCA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05770
Encompass_Preferred
The Human Major Histocompatibility Complex Class I A (MHCA) ELISA Kit allows for quantitative determination of Major Histocompatibility Complex Class I A in Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.126 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P16189. Cross Activity: No significant cross-reactivity or interference between Major Histocompatibility Complex Class I A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3194