Learn More
Biomatik Corporation Human Metastasis Associated In Colon Cancer 1 (MACC1) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU05926
The Human Metastasis Associated In Colon Cancer 1 (MACC1) ELISA Kit allows for quantitative determination of Metastasis Associated In Colon Cancer 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.116 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q6ZN28. Cross Activity: No significant cross-reactivity or interference between Metastasis Associated In Colon Cancer 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3261