Learn More
Biomatik Corporation Human Mothers Against Decapentaplegic Homolog 4 (Smad4) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06008
The Human Mothers Against Decapentaplegic Homolog 4 (Smad4) ELISA Kit allows for quantitative determination of Mothers Against Decapentaplegic Homolog 4 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.113 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q13485. Cross Activity: No significant cross-reactivity or interference between Mothers Against Decapentaplegic Homolog 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3283