Learn More
Biomatik Corporation Mouse Myosin Heavy Chain 2, Skeletal Muscle, Adult (MYH2) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06092
The Mouse Myosin Heavy Chain 2, Skeletal Muscle, Adult (MYH2) ELISA Kit allows for quantitative determination of Myosin Heavy Chain 2, Skeletal Muscle, Adult in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.121 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: I6L963. Cross Activity: No significant cross-reactivity or interference between Myosin Heavy Chain 2, Skeletal Muscle, Adult and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3317