Learn More
Biomatik Corporation Human Myxovirus Resistance 1 (MX1) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06125
The Human Myxovirus Resistance 1 (MX1) ELISA Kit allows for quantitative determination of Myxovirus Resistance 1 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.65 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P20591. Cross Activity: No significant cross-reactivity or interference between Myxovirus Resistance 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3325