Learn More
Biomatik Corporation Human N-Acetylgalactosaminidase Alpha (NAGa) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06132
The Human N-Acetylgalactosaminidase Alpha (NAGa) ELISA Kit allows for quantitative determination of N-Acetylgalactosaminidase Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.134 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P17050. Cross Activity: No significant cross-reactivity or interference between N-Acetylgalactosaminidase Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3329