Learn More
Biomatik Corporation Mouse Neuropeptide FF (NPFF) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06242
The Mouse Neuropeptide FF (NPFF) ELISA Kit allows for quantitative determination of Neuropeptide FF in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.2-500 pg/mL. Sensitivity: 13.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: Q9WVA8. Cross Activity: No significant cross-reactivity or interference between Neuropeptide FF and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3359