Learn More
Biomatik Corporation Rat Osteocalcin (OC) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06416
The Rat Osteocalcin (OC) ELISA Kit allows for quantitative determination of Osteocalcin in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04640. Cross Activity: No significant cross-reactivity or interference between Osteocalcin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3424