Learn More
Biomatik Corporation Human Parathyroid Hormone (PTH) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06482
The Human Parathyroid Hormone (PTH) ELISA Kit allows for quantitative determination of Parathyroid Hormone in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.35 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01270. Cross Activity: No significant cross-reactivity or interference between Parathyroid Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3459