Learn More
Biomatik Corporation Rat Peripheral Myelin Protein 22 (PMP22) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06543
The Rat Peripheral Myelin Protein 22 (PMP22) ELISA Kit allows for quantitative determination of Peripheral Myelin Protein 22 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25094. Cross Activity: No significant cross-reactivity or interference between Peripheral Myelin Protein 22 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3486