Learn More
Biomatik Corporation Mouse Pigment Epithelium Derived Factor (PEDF) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06639
The Mouse Pigment Epithelium Derived Factor (PEDF) ELISA Kit allows for quantitative determination of Pigment Epithelium Derived Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P97298. Cross Activity: No significant cross-reactivity or interference between Pigment Epithelium Derived Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3522