Learn More
Biomatik Corporation Mouse Plasminogen Activator, Urokinase Receptor (uPAR) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06676
The Mouse Plasminogen Activator, Urokinase Receptor (uPAR) ELISA Kit allows for quantitative determination of Plasminogen Activator, Urokinase Receptor in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 78-5,000 pg/mL. Sensitivity: 28 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P35456. Cross Activity: No significant cross-reactivity or interference between Plasminogen Activator, Urokinase Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3540