Learn More
Biomatik Corporation Rat Plasminogen Activator, Urokinase Receptor (uPAR) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06677
The Rat Plasminogen Activator, Urokinase Receptor (uPAR) ELISA Kit allows for quantitative determination of Plasminogen Activator, Urokinase Receptor in Serum, Plasma and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.122 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P49616. Cross Activity: No significant cross-reactivity or interference between Plasminogen Activator, Urokinase Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3541