Learn More
Biomatik Corporation Human Platelet Derived Growth Factor AA (PDGFAA) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06683
The Human Platelet Derived Growth Factor AA (PDGFAA) ELISA Kit allows for quantitative determination of Platelet Derived Growth Factor AA in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.065 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04085. Cross Activity: No significant cross-reactivity or interference between Platelet Derived Growth Factor AA and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3543