Learn More
Biomatik Corporation Rat Platelet Derived Growth Factor D (PDGFD) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06690
The Rat Platelet Derived Growth Factor D (PDGFD) ELISA Kit allows for quantitative determination of Platelet Derived Growth Factor D in Serum, Plasma and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9EQT1. Cross Activity: No significant cross-reactivity or interference between Platelet Derived Growth Factor D and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3548