Learn More
Biomatik Corporation Human Platelet Derived Growth Factor Receptor Alpha (PDGFRa) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06691
The Human Platelet Derived Growth Factor Receptor Alpha (PDGFRa) ELISA Kit allows for quantitative determination of Platelet Derived Growth Factor Receptor Alpha in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.064 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P16234. Cross Activity: No significant cross-reactivity or interference between Platelet Derived Growth Factor Receptor Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3549