Learn More
Biomatik Corporation Human Poliovirus Receptor Related Protein 4 (PVRL4) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06731
The Human Poliovirus Receptor Related Protein 4 (PVRL4) ELISA Kit allows for quantitative determination of Poliovirus Receptor Related Protein 4 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 125-8000 pg/mL. Sensitivity: 52 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96NY8. Cross Activity: No significant cross-reactivity or interference between Poliovirus Receptor Related Protein 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3569