Learn More
Biomatik Corporation Human Procollagen C Proteinase Enhancer 1 (PCPE1) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06768
The Human Procollagen C Proteinase Enhancer 1 (PCPE1) ELISA Kit allows for quantitative determination of Procollagen C Proteinase Enhancer 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.285 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q15113. Cross Activity: No significant cross-reactivity or interference between Procollagen C Proteinase Enhancer 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3581