Learn More
Biomatik Corporation Human Procollagen II N-Terminal Propeptide (PIINP) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06781
The Human Procollagen II N-Terminal Propeptide (PIINP) ELISA Kit allows for quantitative determination of Procollagen II N-Terminal Propeptide in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 93.75-6,000 pg/mL. Sensitivity: 37.55 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02458. Cross Activity: No significant cross-reactivity or interference between Procollagen II N-Terminal Propeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3589