Learn More
Biomatik Corporation Human Procollagen III N-Terminal Propeptide (PIIINP) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06786
The Human Procollagen III N-Terminal Propeptide (PIIINP) ELISA Kit allows for quantitative determination of Procollagen III N-Terminal Propeptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.25-400 ng/mL. Sensitivity: 2.14 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02461. Cross Activity: No significant cross-reactivity or interference between Procollagen III N-Terminal Propeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3590