Learn More
Biomatik Corporation Mouse Proenkephalin (PENK) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU06793
The Mouse Proenkephalin (PENK) ELISA Kit allows for quantitative determination of Proenkephalin in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 78-5000 pg/mL. Sensitivity: 29 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P22005. Cross Activity: No significant cross-reactivity or interference between Proenkephalin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3595